Tisk…

Dihydrogenfosforečnan draselný, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

M-Clarity™ quality level = MQ200

P5655-100G P5655-1KG P5655-500G
SIALP5655-100GEA 1240 CZK
SIALP5655-100G SIALP5655-1KG SIALP5655-500G
Dihydrogenfosforečnan draselný, Sigma-Aldrich®
Dihydrogenfosforečnan draselný
Vzorec: KH₂PO₄
Bod varu: ∼100 °C (1013 hPa)
Bod tání: 253 °C
Hustota: 2,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS číslo: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR