Tisk…

Hydrogenfosforečnan dvojsodný, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

M-Clarity™ quality level = MQ200

S3264-1KG S3264-250G S3264-500G
SIALS3264-1KGEA 11940 CZK
SIALS3264-1KG SIALS3264-250G SIALS3264-500G
Hydrogenfosforečnan dvojsodný, Sigma-Aldrich®
Hydrogenfosforečnan dvojsodný
Vzorec: Na₂HPO₄
MW: 141.96
Bod tání: 250
Hustota: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS číslo: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR