, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
60353-1KG
60353-250G
SIAL60353-1KGEA
10340
CZK
SIAL60353-1KG
SIAL60353-250G
, Sigma-Aldrich®
Hydrogenfosforečnan didraselný
Vzorec:
K₂HPO₄ MW: 174.18 Bod tání: 340 Hustota: 2.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011383 CAS číslo: 7758-11-4 EINECS: 231-834-5 Merck Index: 13,07743 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...