Tisk…

Potassium hexafluorophosphate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

515973-25G 515973-5G
SIAL515973-25GEA 5580 CZK
SIAL515973-25G SIAL515973-5G
Potassium hexafluorophosphate, Sigma-Aldrich®
Potassium hexafluorophosphate
Vzorec: F₆KP
Bod tání: 575 °C
Hustota: 2,58 g/cm³ (20 °C)
MDL Number: MFCD00011412
CAS číslo: 17084-13-8
EINECS: 241-143-0
UN: 3260
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR