Tisk…

Potassium hexafluorophosphate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A12080.0B A12080.18 A12080.30
A12080.0BEA 11950 CZK
A12080.0B A12080.18 A12080.30
Potassium hexafluorophosphate ≥98%
Potassium hexafluorophosphate

Purity: 98% CAS No.: 17084-13-8 Molecular Formula: KPF6 Formula Weight: 184.07 Odour: Odourless pH: 3.0-7.0 Melting Point: 575° UN No.: 3260 MDL No.: MFCD00011412

Vzorec: F₆KP
Bod tání: 575 °C
Hustota: 2,55
Storage Temperature: Ambient
MDL Number: MFCD00011412
CAS číslo: 17084-13-8
EINECS: 241-143-0
UN: 3260
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR