Tisk…

Tetrachlorpaladnatan dvojsodný ≥98%, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Palladium sodium chloride, Sodium palladium (II) chloride, Disodium tetrachloropalladate

M-Clarity™ quality level = MQ100

205818-1G 205818-25G 205818-5G
SIAL205818-1GEA 3510 CZK
SIAL205818-1G SIAL205818-25G SIAL205818-5G
Tetrachlorpaladnatan dvojsodný ≥98%, Sigma-Aldrich®
Tetrachlorpaladnatan dvojsodný
Vzorec: Cl₄Na₂Pd
MW: 294,21 g/mol
MDL Number: MFCD00003487
CAS číslo: 13820-53-6
EINECS: 237-502-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR