Sodium cyanate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
185086-100G
185086-500G
185086-5G
SIAL185086-100GEA
1010
CZK
SIAL185086-100G
SIAL185086-500G
SIAL185086-5G
Sodium cyanate, Sigma-Aldrich®
Sodium cyanate
Vzorec:
CNNaO Bod tání: 550 °C Hustota: 1,893 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011121 CAS číslo: 917-61-3 EINECS: 213-030-6 Merck Index: 12,08749 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...