Tisk…

Potassium superoxide, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Potassium dioxide

M-Clarity™ quality level = MQ100

278904-250G 278904-50G
SIAL278904-250GEA 6500 CZK
SIAL278904-250G SIAL278904-50G
Potassium superoxide, Sigma-Aldrich®
Potassium superoxide
Vzorec: KO₂
Bod tání: 560 °C
Hustota: 2,14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082411
CAS číslo: 12030-88-5
EINECS: 234-746-5
UN: 2466
ADR: 5.1,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR