Tisk…

Sodium dicyanoamide, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Sodium dicyanamide

M-Clarity™ quality level = MQ100

690309-10KG 690309-1KG 690309-80KG
SIAL690309-10KGEA 0 CZK
SIAL690309-10KG SIAL690309-1KG SIAL690309-80KG
Sodium dicyanoamide, Sigma-Aldrich®
Sodium dicyanoamide
Vzorec: C₂N₃Na
MW: 89.03
Storage Temperature: Ambient
MDL Number: MFCD00003535
CAS číslo: 1934-75-4
EINECS: 217-703-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR