Jodid rubidny ≥99.8% (metals basis)
Dodavatel: Thermo Fisher Scientific
13497.09
13497.18
13497.A7
13497.09EA
2750
CZK
13497.09
13497.18
13497.A7
Jodid rubidny ≥99.8% (metals basis)
Jodid rubidny
Purity: 99.8%
CAS No.: 7790-29-6
Molecular Formula: RbI
Formula Weight: 212.37
Physical Form: Crystalline
Density: 3.55
Refractive Index: 1.6474
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 647°
Boiling Point: 1300°
Solubility: Freely soluble in water. Soluble in alcohol
Merck Reference: 14,8286
MDL No.: MFCD00011192
Vzorec:
IRb Bod varu: 1300 °C Bod tání: 647 °C Hustota: 3,55 Storage Temperature: Ambient |
MDL Number:
MFCD00011192 CAS číslo: 7790-29-6 EINECS: 232-198-1 Merck Index: 14,8286 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...