Jodid lithný, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
450952-25G
450952-5G
SIAL450952-25GEA
23070
CZK
SIAL450952-25G
SIAL450952-5G
Jodid lithný, Sigma-Aldrich®
Jodid lithný
Vzorec:
ILi MW: 133.85 Bod varu: 1177 Bod tání: 450 Hustota: 3.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011092 CAS číslo: 10377-51-2 EINECS: 233-822-5 Merck Index: 13,05557 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...