Hydrid sodný, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
223441-50G
223441-10G
223441-1KG
SIAL223441-50GEA
7620
CZK
SIAL223441-50G
SIAL223441-10G
SIAL223441-1KG
Hydrid sodný, Sigma-Aldrich®
Hydrid sodný
Vzorec:
HNa MW: 24 Storage Temperature: Ambient |
MDL Number:
MFCD00003471 CAS číslo: 7646-69-7 EINECS: 231-587-3 UN: 1427 ADR: 4.3 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...