Lithium hydride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
201049-100G
201049-10G
201049-500G
201049-5G
SIAL201049-100GEA
6520
CZK
SIAL201049-100G
SIAL201049-10G
SIAL201049-500G
SIAL201049-5G
Lithium hydride, Sigma-Aldrich®
Lithium hydride
Vzorec:
HLi MW: 7,95 g/mol Bod tání: 680 °C Hustota: 0,685…0.725 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011074 CAS číslo: 7580-67-8 EINECS: 231-484-3 UN: 1414 ADR: 4.3,I Merck Index: 13,05555 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...