Tisk…

Hexakyanoželezitan draselný ≥99.0% ACS

Dodavatel: Thermo Scientific
Varování

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

33357.36 33357.A3
33357.36EA 2670 CZK
33357.36 33357.A3
Hexakyanoželezitan draselný ≥99.0% ACS
Hexakyanoželezitan draselný

Purity: 99.0% min CAS No.: 13746-66-2 Molecular Formula: K3Fe(CN)6 Formula Weight: 329.26 Physical Form: Crystalline Density: 1.89 Melting Point: dec. Merck Reference: 14,7630 MDL No.: MFCD00011392

Vzorec: K₃Fe(CN)₆
MW: 329,25 g/mol
Bod tání: Dec.
Hustota: 1,89
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS číslo: 13746-66-2
EINECS: 237-323-3
Merck Index: 14,7630

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR