Tisk…

Rubidium fluoride 99.975% (metals basis), Puratronic®

Dodavatel: Thermo Scientific
Nebezpečí

12873.04 12873.09
12873.04EA 3540 CZK
12873.04 12873.09
Rubidium fluoride 99.975% (metals basis), Puratronic®
Rubidium fluoride

Purity: 99.975% CAS No.: 13446-74-7 Molecular Formula: FRb Formula Weight: 104.47 Physical Form: Crystalline Density: 3.557 Refractive Index: 1.398 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 795° UN No.: 3288 MDL No.: MFCD00011191

Vzorec: FRb
Bod varu: 1408 °C (1013 hPa)
Bod tání: 795 °C
Hustota: 3,557
Storage Temperature: Ambient
MDL Number: MFCD00011191
CAS číslo: 13446-74-7
EINECS: 236-603-2
UN: 3288
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR