Potassium fluoride dihydrate ≥98.5%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
11555.14
11555.30
11555.A1
11555.14EA
646
CZK
11555.14
11555.30
11555.A1
Potassium fluoride dihydrate ≥98.5%
Potassium fluoride dihydrate
Purity: 98.5+%
CAS No.: 13455-21-5
Molecular Formula: KF.2H2O
Formula Weight: 94.13 (58.10anhy)
Physical Form: Crystalline
Density: 2.5
Refractive Index: 1.352
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 41°
Solubility: Freely soluble in water
Merck Reference: 14,7632
UN No.: 1812
MDL No.: MFCD00149929
Vzorec:
FK·2H₂O MW: 94.13 Bod varu: 1505 Bod tání: 41 °C Hustota: 2.5 Storage Temperature: Ambient |
MDL Number:
MFCD00149929 CAS číslo: 13455-21-5 EINECS: 232-151-5 UN: 1812 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...