Tisk…

Bromid cesný ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Cesiumbromide

12928.18 12928.30
12928.18EA 3330 CZK
12928.18 12928.30
Bromid cesný ≥99.9% (metals basis)
Bromid cesný

Purity: 99.9% CAS No.: 7787-69-1 Molecular Formula: BrCs Formula Weight: 212.81 Physical Form: Crystalline Density: 4.44 Refractive Index: 1.6984 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 636° Boiling Point: 1300° Merck Reference: 14,2009 MDL No.: MFCD00010954

Vzorec: BrCs
MW: 212.81
Bod varu: 1300 °C
Bod tání: 636
Hustota: 4.44
Storage Temperature: Ambient
MDL Number: MFCD00010954
CAS číslo: 7787-69-1
EINECS: 232-130-0
Merck Index: 14,2009

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR