Tisk…

Tetrafenylboritan sodný ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

A10909.14 A10909.06 A10909.22
A10909.14EA 4840 CZK
A10909.14 A10909.06 A10909.22
Tetrafenylboritan sodný ≥99%
Tetrafenylboritan sodný

Purity: 99% CAS No.: 143-66-8 Molecular Formula: C24H20BNa Formula Weight: 342.23 pH: 8 Melting Point: 300° Beilstein Reference: 3599783 Merck Reference: 14,8690 UN No.: 2811 MDL No.: MFCD00011494

Vzorec: (C₆H₅)₄BNa
MW: 342.22
Bod tání: >300 °C
Hustota: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS číslo: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,08765

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR