Tisk…

Potassium tetrafluoroborate ≥98%

Dodavatel: Thermo Scientific
Varování

A15605.30 A15605.0B A15605.0I
A15605.30EA 1210 CZK
A15605.30 A15605.0B A15605.0I
Potassium tetrafluoroborate ≥98%
Potassium tetrafluoroborate

Purity: 98% CAS No.: 14075-53-7 Molecular Formula: BF4K Formula Weight: 125.91 Refractive Index: 1.3245 Melting Point: 529° Boiling Point: dec. Merck Reference: 14,7686 MDL No.: MFCD00011395

Vzorec: BF₄K
MW: 125,9 g/mol
Bod varu: Dec.
Bod tání: 529 °C
Hustota: 2,50
Storage Temperature: Ambient
MDL Number: MFCD00011395
CAS číslo: 14075-53-7
EINECS: 237-928-2
Merck Index: 14,7686

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR