Potassium tetrafluoroborate ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
11556.36
11556.A3
11556.36EA
2250
CZK
11556.36
11556.A3
Potassium tetrafluoroborate ≥99%
Potassium tetrafluoroborate
Purity: 99% min
CAS No.: 14075-53-7
Molecular Formula: BF4K
Formula Weight: 125.91
Physical Form: Powder
Density: 2.50
Refractive Index: 1.3245
Sensitivity: Hygroscopic
Melting Point: 529°
Boiling Point: dec.
Solubility: Soluble in water (0.45g/100g H2O). Slightly soluble in boiling alcohol
Merck Reference: 14,7686
MDL No.: MFCD00011395
Vzorec:
BF₄K MW: 125,9 g/mol Bod varu: Dec. Bod tání: 529 °C Hustota: 2,50 Storage Temperature: Ambient |
MDL Number:
MFCD00011395 CAS číslo: 14075-53-7 EINECS: 237-928-2 Merck Index: 14,7686 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...