Sodium tetraethylborate
Dodavatel: LGC Standards PROMOCHEM
|
Nebezpečí
|
DE-MS-03401-1
DE-MS-03401-25
DE-MS-03401-5
PROODE-MS-03401-1EA
4270
CZK
PROODE-MS-03401-1
PROODE-MS-03401-25
PROODE-MS-03401-5
Sodium tetraethylborate
Sodium tetraethylborate
Vzorec:
C₈H₂₀BNa |
MDL Number:
MFCD00061547 CAS číslo: 15523-24-7 UN: 3393 ADR: 4.2,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...