Tisk…

Methylphosphonic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Methanephosphonic acid

A12619.03 A12619.06 A12619.14 A12619.22
A12619.03EA 1280 CZK
A12619.03 A12619.06 A12619.14 A12619.22
Methylphosphonic acid ≥98%
Methylphosphonic acid

Purity: 98% CAS No.: 993-13-5 Molecular Formula: CH5O3P Formula Weight: 96.03 Melting Point: 103-107° Beilstein Reference: 1739372 UN No.: 3261 MDL No.: MFCD00002137

Vzorec: CH₅O₃P
Bod tání: 103…107 °C
Hustota: 1,639 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002137
CAS číslo: 993-13-5
EINECS: 213-607-2
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR