Tisk…

Voda for molecular biology nuclease-free, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Hydrogen oxide

M-Clarity™ quality level = MQ200

W4502-1L W4502-10L W4502-6X1L
SIALW4502-1LEA 4300 CZK
SIALW4502-1L SIALW4502-10L SIALW4502-6X1L
Voda for molecular biology nuclease-free, Sigma-Aldrich®
Voda
Vzorec: H₂O
Bod varu: 100 °C (1013 hPa)
Bod tání: 0 °C
Hustota: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS číslo: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR