Voda, PCR grade, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Hydrogen oxide
MQ200
W1754-5VL
W1754-1VL
SIALW1754-5VLEA
7800
CZK
SIALW1754-5VL
SIALW1754-1VL
Voda, PCR grade, Sigma-Aldrich®
Voda
PCR grade water is sterile-filtered. It is free from exonucleases (DNAse, RNAse) and endonuclease (NICKase) and is also free from nucleic acid contamination.
Vzorec:
H₂O MW: 18.02 Bod varu: 100 Bod tání: 0 Hustota: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS číslo: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...