Voda, deuterium depleted, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Hydrogen oxide
M-Clarity™ quality level = MQ200
195294-100G
195294-10G
195294-25G
SIAL195294-100GEA
9080
CZK
SIAL195294-100G
SIAL195294-10G
SIAL195294-25G
Voda, deuterium depleted, Sigma-Aldrich®
Voda
Vzorec:
H₂O MW: 18.02 Bod varu: 100 Bod tání: 0 Hustota: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS číslo: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...