Tisk…

Voda, UltraPure, sterile filtered

Dodavatel: Rockland Immunochemicals

Synonyms: Hydrogen oxide

MB-009-1000 MB-009-4000
ROCKMB-009-1000EA 3490 CZK
ROCKMB-009-1000 ROCKMB-009-4000
Voda, UltraPure, sterile filtered
Voda
Vzorec: H₂O
Bod varu: 100 °C (1013 hPa)
Bod tání: 0 °C
Hustota: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS číslo: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR