Tisk…
Kyanoguanidin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Dicyanodiamide, 1-Cyanoguanidine, N-Cyanoguanidine, Dicyandiamide
M-Clarity™ quality level = MQ200
D76609-1KG D76609-25G D76609-3KG
SIALD76609-1KGEA 1450 CZK
SIALD76609-1KG SIALD76609-25G SIALD76609-3KG
Kyanoguanidin, Sigma-Aldrich®
Kyanoguanidin
Vzorec: (H₂N)₂CNCN
MW: 84.08
Bod varu: 143
Hustota: 1.404 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008066
CAS číslo: 461-58-5
EINECS: 207-312-8
Merck Index: 13,03119