Tisk…

Kyanoguanidin for synthesis, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dicyanodiamide, 1-Cyanoguanidine, N-Cyanoguanidine, Dicyandiamide

M-Clarity™ quality level = MQ200

8.02491.0100 8.02491.1000
8.02491.0100EA 821 CZK
8.02491.0100 8.02491.1000
Kyanoguanidin for synthesis, Sigma-Aldrich®
Kyanoguanidin
Vzorec: (H₂N)₂CNCN
Bod varu: 143 °C (1013 hPa)
Hustota: 1,404 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008066
CAS číslo: 461-58-5
EINECS: 207-312-8
Merck Index: 13,03119

Objednat

Výsledky specifikačního testu

Solubility 34.6 g/l (20°C)
Melting Point 208 - 210 °C
Molar Mass 400 kg/m3
Bulk Density 400 kg/m3
Density 1.4 g/cm3 (20 °C)
Synonyms Dicyanodiamide
Hill Formula C2H4N4
Chemical Formula NCNHC(=NH)NH2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR