Kyanoguanidin ≥99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Dicyanodiamide, 1-Cyanoguanidine, N-Cyanoguanidine, Dicyandiamide
A10451.0E
A10451.30
A10451.36
A10451.0EEA
3320
CZK
A10451.0E
A10451.30
A10451.36
Kyanoguanidin ≥99%
Kyanoguanidin
Purity: 99%
CAS No.: 461-58-5
Molecular Formula: C2H4N4
Formula Weight: 84.08
Physical Form: Powder
Density: 1.400
Appearance: White
Odour: Odourless
Melting Point: 208-211°
Beilstein Reference: 605637
Merck Reference: 14,3092
MDL No.: MFCD00008066
Vzorec:
(H₂N)₂CNCN MW: 84.08 Bod varu: 143 Bod tání: 208…211 °C Hustota: 1.400 Storage Temperature: Ambient |
MDL Number:
MFCD00008066 CAS číslo: 461-58-5 EINECS: 207-312-8 Merck Index: 13,03119 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...