Tisk…

Kyanoguanidin 99.5%

Dodavatel: Thermo Fisher Scientific

Synonyms: Dicyanodiamide, 1-Cyanoguanidine, N-Cyanoguanidine, Dicyandiamide

113810010 113810025 113810050 113810250
ACRO113810010EA 1800 CZK
ACRO113810010 ACRO113810025 ACRO113810050 ACRO113810250
Kyanoguanidin 99.5%
Kyanoguanidin
Vzorec: (H₂N)₂CNCN
MW: 84.08
Bod varu: 143
Bod tání: 208…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00008066
CAS číslo: 461-58-5
EINECS: 207-312-8
Merck Index: 13,03119

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 208 to 211 °C
Assay ≥99.4 % (ex N)
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR