Tisk…

2-Methoxyfuran ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15593.03 A15593.06
A15593.03EA 2700 CZK
A15593.03 A15593.06
2-Methoxyfuran ≥97%
2-Methoxyfuran

Purity: 97% CAS No.: 25414-22-6 Molecular Formula: C5H6O2 Formula Weight: 98.10 Storage Temperature: 2°C to 8°C Flash Point: 10°(50°F) Refractive Index: 1.4470 Boiling Point: 110-111° Beilstein Reference: 106270 UN No.: 3271 MDL No.: MFCD00003227

Vzorec: C₅H₆O₂
Bod varu: 110…111 °C
Hustota: 1,065
Bod vzplanutí: 10 °C (50 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003227
CAS číslo: 25414-22-6
EINECS: 246-953-8
UN: 3271
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR