5-Iodo-2-furaldehyde ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-Iodofurfural
L19670.03
L19670.06
L19670.03EA
1390
CZK
L19670.03
L19670.06
5-Iodo-2-furaldehyde ≥97%
5-Iodo-2-furaldehyde
Purity: 97%
CAS No.: 2689-65-8
Molecular Formula: C5H3IO2
Formula Weight: 221.98
Storage Temperature: 2°C to 8°C
Melting Point: 126-127°
Beilstein Reference: 108406
MDL No.: MFCD00159503
Vzorec:
C₅H₃IO₂ MW: 221,98 g/mol Bod tání: 126…127 °C |
MDL Number:
MFCD00159503 CAS číslo: 2689-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...