5-Nitro-2-furaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
5-Nitrofurfural
M-Clarity™ quality level = MQ100
170968-25G
170968-5G
SIAL170968-25GEA
4410
CZK
SIAL170968-25G
SIAL170968-5G
5-Nitro-2-furaldehyde, Sigma-Aldrich®
5-Nitro-2-furaldehyde
Vzorec:
C₅H₃NO₄ MW: 141,08 g/mol Bod varu: 272 °C (1013 hPa) Bod tání: 35 °C Hustota: 1,34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003230 CAS číslo: 698-63-5 EINECS: 211-816-3 UN: 1325 ADR: 4.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...