Tisk…

5-Bromo-2-furaldehyde ≥98% stabilizovaný

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 5-Bromofuran-2-carbaldehyde, 5-Bromofurfural

Stabilization: Stabilised with ethanol

B22536.09 B22536.18
B22536.09EA 2770 CZK
B22536.09 B22536.18
5-Bromo-2-furaldehyde ≥98% stabilizovaný
5-Bromo-2-furaldehyde

Stabilized with 2% ethanol


Purity: 98% CAS No.: 1899-24-7 Molecular Formula: C5H3BrO2 Formula Weight: 174.98 Melting Point: 82-85° Boiling Point: 112°/6mm MDL No.: MFCD00159501

Vzorec: C₅H₃BrO₂
MW: 174.98
Bod varu: 112 °C (6 mmHg)
Bod tání: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00159501
CAS číslo: 1899-24-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR