Tisk…

Fluorescein dvojsodná sůl, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate

M-Clarity™ quality level = MQ200

F6377-100G
SIALF6377-100GEA 2500 CZK
SIALF6377-100G
Fluorescein dvojsodná sůl, Sigma-Aldrich®
Fluorescein dvojsodná sůl
Vzorec: C₂₀H₁₀Na₂O₅
Bod tání: > 300 °C
Hustota: 1,601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS číslo: 518-47-8
EINECS: 208-253-0
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR