Tisk…

Fluorescein, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid

M-Clarity™ quality level = MQ100

46955-100G-F 46955-500G-F
SIAL46955-100G-FEA 44310 CZK
SIAL46955-100G-F SIAL46955-500G-F
Fluorescein, Sigma-Aldrich®
Fluorescein
Vzorec: C₂₀H₁₂O₅
MW: 332.31
Bod tání: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00005050
CAS číslo: 2321-07-5
EINECS: 219-031-8
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97b9d8d37f0106f4c8ef2d7759759d21bd43cc03&vid=25b78df95a04ab26c5ba8c96b95f031056926b13&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b0e97bec9c9f44dd58e1cd0ca79d7e94c3ba82d&vid=a29b350acb5fd71b32a30baa408af54494e42d59&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR