Fluorescein, pure
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
119241000
119245000
119240025
119240250
ACRO119241000EA
2140
CZK
ACRO119241000
ACRO119245000
ACRO119240025
ACRO119240250
Fluorescein, pure
Fluorescein
Vzorec:
C₂₀H₁₂O₅ Bod tání: 314…316 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005050 CAS číslo: 2321-07-5 EINECS: 219-031-8 Merck Index: 13,04185 |
|
Výsledky specifikačního testu
Appearance | Red-brown to rust brown to brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤3 % |
Sulfated ash | ≤0.5 % |
Specific absorbance A (1%/1cm) | ≥1800 (490 +/- 5 nm , 0.5% aq. soln. of Na2CO3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...