9-Fluorenylmethanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol
M-Clarity™ quality level = MQ200
160504-10G
160504-50G
SIAL160504-10GEA
3350
CZK
SIAL160504-10G
SIAL160504-50G
9-Fluorenylmethanol, Sigma-Aldrich®
9-Fluorenylmethanol
Vzorec:
C₁₄H₁₂O MW: 196.25 Bod varu: 337 Bod tání: 107 |
MDL Number:
MFCD00001139 CAS číslo: 24324-17-2 EINECS: 246-167-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...