Tisk…

9-Fluorenylmethanol ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: (Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol

A15212.06 A15212.14 A15212.22
A15212.06EA 1470 CZK
A15212.06 A15212.14 A15212.22
9-Fluorenylmethanol ≥99%
9-Fluorenylmethanol

Purity: 99% CAS No.: 24324-17-2 Molecular Formula: C14H12O Formula Weight: 196.25 Melting Point: 102-106° Beilstein Reference: 2330017

Vzorec: C₁₄H₁₂O
MW: 196.25
Bod varu: 337
Bod tání: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00001139
CAS číslo: 24324-17-2
EINECS: 246-167-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR