2-Aminofluorene 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
9H-Fluoren-2-amine, 2-Fluorenamine, Fluoren-2-ylamine
B22769.03
B22769.06
B22769.14
B22769.03EA
749
CZK
B22769.03
B22769.06
B22769.14
2-Aminofluorene 98%
2-Aminofluorene
Purity: 98%
CAS No.: 153-78-6
Molecular Formula: C13H11N
Formula Weight: 181.24
Melting Point: 126-130°
Beilstein Reference: 1945861
Vzorec:
C₁₃H₁₁N MW: 181,24 g/mol Bod varu: 327 °C (1013 hPa) Bod tání: 126…130 °C Hustota: 1,281 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001125 CAS číslo: 153-78-6 EINECS: 205-817-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...