9,10-Phenanthrenequinone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Phenanthrene-9,10-dione
M-Clarity™ quality level = MQ100
275034-100G
275034-25G
SIAL275034-100GEA
12640
CZK
SIAL275034-100G
SIAL275034-25G
9,10-Phenanthrenequinone, Sigma-Aldrich®
9,10-Phenanthrenequinone
Vzorec:
C₁₄H₈O₂ MW: 208,22 g/mol Bod varu: 360 °C (1013 hPa) Bod tání: 203…206 °C Hustota: 1,4045 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001163 CAS číslo: 84-11-7 EINECS: 201-515-5 UN: 3077 ADR: 9,III Merck Index: 12,07355 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...