9-Fluorenon, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
9H-Fluoren-9-one
M-Clarity™ quality level = MQ200
F1506-100G-A
F1506-500G-A
F1506-5G-A
SIALF1506-100G-AEA
1380
CZK
SIALF1506-100G-A
SIALF1506-500G-A
SIALF1506-5G-A
9-Fluorenon, Sigma-Aldrich®
9-Fluorenon
Vzorec:
C₁₃H₈O MW: 180.21 Bod varu: 342 Bod tání: 81…83 °C Hustota: 0.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001141 CAS číslo: 486-25-9 EINECS: 207-630-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...