Tisk…

1-Ethylpiperazine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-Ethylpiperazine

415308-50ML 415308-250ML
SIAL415308-50MLEA 1320 CZK
SIAL415308-50ML SIAL415308-250ML
1-Ethylpiperazine, Sigma-Aldrich®
1-Ethylpiperazine
Vzorec: C₆H₁₄N₂
MW: 114.19
Bod varu: 157
Bod tání: –60 °C
Hustota: 0.8989
Storage Temperature: Ambient
MDL Number: MFCD00059912
CAS číslo: 5308-25-8
EINECS: 226-166-6
UN: 2733
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR