Hexaphenylcyclotrisiloxane ≥98%
Dodavatel: Thermo Fisher Scientific
B23061.06
B23061.14
B23061.06EA
2960
CZK
B23061.06
B23061.14
Hexaphenylcyclotrisiloxane ≥98%
Hexaphenylcyclotrisiloxane
Purity: 98+%
CAS No.: 512-63-0
Molecular Formula: C36H30O3Si3
Formula Weight: 594.90
Melting Point: ca 175-185°
Boiling Point: 300°/1mm
MDL No.: MFCD00023110
Vzorec:
C₃₆H₃₀O₃Si₃ Bod varu: 300 °C (1 mmHg) Bod tání: ∼ 175…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023110 CAS číslo: 512-63-0 EINECS: 208-145-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...