Tisk…

4,4-Dimethyl-2-cyclohexen-1-one, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4,4-Dimethylcyclohex-2-enone

M-Clarity™ quality level = MQ200

245542-25G 245542-5G
SIAL245542-25GEA 0 CZK
SIAL245542-25G SIAL245542-5G
4,4-Dimethyl-2-cyclohexen-1-one, Sigma-Aldrich®
4,4-Dimethyl-2-cyclohexen-1-one
Vzorec: C₈H₁₂O
MW: 124,18 g/mol
Bod varu: 27,5 °C (0.2 mmHg)
Hustota: 0,93…0.935 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009695
CAS číslo: 1073-13-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR