3-Methyl-2-cyclopenten-1-one, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone
M-Clarity™ quality level = MQ300
W343502-100G
W343502-25G
W343502-SAMPLE
SIALW343502-100GEA
35270
CZK
SIALW343502-100G
SIALW343502-25G
SIALW343502-SAMPLE
3-Methyl-2-cyclopenten-1-one, Sigma-Aldrich®
3-Methyl-2-cyclopenten-1-one
Vzorec:
C₆H₈O MW: 96,13 g/mol Bod varu: 158 °C (1013 hPa) Bod tání: 2 °C Hustota: 0,97 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001403 CAS číslo: 2758-18-1 EINECS: 220-421-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...