Tisk…

2-Cyclohexen-1-ol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cyclohex-2-ene-1-ol, Cyclohex-2-enol

M-Clarity™ quality level = MQ100

C102652-10G C102652-1G C102652-5G
SIALC102652-10GEA 12870 CZK
SIALC102652-10G SIALC102652-1G SIALC102652-5G
2-Cyclohexen-1-ol, Sigma-Aldrich®
2-Cyclohexen-1-ol
Vzorec: C₆H₁₀O
MW: 98,14 g/mol
Bod varu: 164…165 °C (1013 hPa)
Hustota: 0,9865 g/cm³ (23 °C)
MDL Number: MFCD00001570
CAS číslo: 822-67-3
EINECS: 212-501-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR