Tisk…

2-Cyclohexen-1-one, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

C102814-100ML C102814-10ML C102814-25ML
SIALC102814-100MLEA 3920 CZK
SIALC102814-100ML SIALC102814-10ML SIALC102814-25ML
2-Cyclohexen-1-one, Sigma-Aldrich®
2-Cyclohexen-1-one
Vzorec: C₆H₈O
MW: 96,13 g/mol
Bod varu: 168 °C (1013 hPa)
Hustota: 0,99 g/cm³ (20 °C)
MDL Number: MFCD00001577
CAS číslo: 930-68-7
EINECS: 213-223-5
UN: 2929
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR