Tisk…

α-Tropolone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

T89702-1G T89702-25G T89702-5G
SIALT89702-1GEA 1540 CZK
SIALT89702-1G SIALT89702-25G SIALT89702-5G
α-Tropolone, Sigma-Aldrich®
α-Tropolone
Vzorec: C₇H₆O₂
Bod varu: 66…67 °C (0,1 mmHg)
Bod tání: 48…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004158
CAS číslo: 533-75-5
EINECS: 208-577-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR