Perfluoro-1,2-dimethylcyclobutane ≥97% remainder 1,3-isomer
Dodavatel: Thermo Fisher Scientific
Synonyms:
1,2-Di(trifluoromethyl)hexafluorocyclobutane, Perfluoro-1,2-dimethylcyclobutane, 1,2-Dimethylperfluorocyclobutane
L16757.14
L16757.22
L16757.14EA
2140
CZK
L16757.14
L16757.22
Perfluoro-1,2-dimethylcyclobutane ≥97% remainder 1,3-isomer
Perfluoro-1,2-dimethylcyclobutane
Purity: 97%
CAS No.: 2994-71-0
Molecular Formula: C6F12
Formula Weight: 300.05
Refractive Index: 1.2540
Melting Point: -32°
Boiling Point: 45°
MDL No.: MFCD00013737
Vzorec:
C₆F₁₂ Bod varu: 45 °C Bod tání: –32 °C Hustota: 1,620 Storage Temperature: Ambient |
MDL Number:
MFCD00013737 CAS číslo: 2994-71-0 EINECS: 221-065-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...